Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027764.2 | NP_082040.2 | 10 | 1596 | ||
GenBank mRNA | AK033501.1 | - | 10 | 1530 | ||
AK088299.1 | - | 10 | 1585 | |||
AK089923.1 | - | 11 | 1597 | |||
AK187668.1 | - | 3 | 179 | |||
BC040764.1 | - | 3 | 291 | |||
BC067005.1 | - | 11 | 1650 | 5721370 |