Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430402E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027768.3 | NP_082044.1 | 1 | 123 | ||
GenBank mRNA | AK017258.1 | - | 1 | 124 |
Gene Symbol: Gm14744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085544.1 | NP_001079013.1 | 1 | 123 |