Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290521.1 | NP_001277450.1 | 4 | 436 | ||
NM_001290522.1 | NP_001277451.1 | 3, 4 | 326 | |||
NM_023597.2 | NP_076086.1 | 4 | 329 | |||
GenBank mRNA | AF274321.1 | - | 4 | 306 | ||
AK017259.1 | - | 3, 4 | 319 | |||
AK141026.1 | - | 4 | 434 | |||
BC026500.1 | - | 3 | 215 | |||
BC037102.1 | - | 4 | 319 | 5364307 | ||
CK618879.1 | - | 4 | 436 |