Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183186.2 | NP_899009.2 | 2, 3 | 609 | ||
GenBank mRNA | AK017346.1 | - | 1, 2 | 412 | ||
AK046748.1 | - | 2, 3 | 884 | |||
BC099971.1 | - | 2, 3 | 687 | 30008715 | ||
BY717160.1 | - | 1, 2 | 407 |