Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5430419D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175166.3 | NP_780375.2 | 18, 19 | 1670 | ||
GenBank mRNA | AK035662.1 | - | 18, 19 | 1671 |