Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001609.3 | NP_001600.1 | 3 | 370 | ||
GenBank mRNA | AK314241.1 | - | 3 | 266 | ||
AL831821.2 | - | 3 | 239 | |||
BC013756.1 | - | 3 | 265 | 4706281 | ||
DA964130.1 | - | 3 | 372 | |||
KU177879.1 | - | 3 | 253 | |||
KU177880.1 | - | 4 | 561 | |||
U12778.1 | - | 3 | 268 |