Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002439.4 | NP_002430.3 | 13 | 1866 | ||
GenBank mRNA | AK289856.1 | - | 14 | 1835 | ||
BC004177.2 | - | 14 | 1807 | |||
BC011817.2 | - | 14 | 1807 | |||
BC017273.2 | - | 13 | 1789 | |||
BC098436.1 | - | 14 | 1807 | |||
BC130434.1 | - | 13 | 1866 | 40146465 | ||
BC130436.1 | - | 13 | 1866 | 40146467 | ||
J04810.1 | - | 13 | 1847 | |||
U61981.1 | - | 14 | 1775 |