Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168371.2 | NP_001161843.1 | 24 | 3792 | ||
NM_001168372.2 | NP_001161844.1 | 24 | 3789 | |||
NM_001168373.2 | NP_001161845.1 | 24 | 3786 | |||
NM_027804.4 | NP_082080.3 | 24 | 3789 | |||
NM_145407.3 | NP_663382.2 | 23 | 3717 | |||
GenBank mRNA | AK043088.1 | - | 23 | 3553 | ||
AK122396.2 | - | 23 | 3575 | |||
AK146504.1 | - | 24 | 3615 | |||
AK148412.1 | - | 19 | 2777 | |||
AK159473.1 | - | 24 | 3611 | |||
AK160807.1 | - | 24 | 3615 | |||
AK171942.1 | - | 24 | 3618 | |||
BC006916.1 | - | 16 | 2323 | |||
BC046824.1 | - | 15 | 2252 | |||
BC060613.1 | - | 24 | 3717 | 6400986 |