Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027810.3 | NP_082086.2 | 8 | 849 | ||
GenBank mRNA | AF521645.1 | - | 8 | 848 | ||
AK050291.1 | - | 8 | 847 | |||
AK132661.1 | - | 8 | 777 | |||
BC031505.1 | - | 8 | 799 | 3494587 |