Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kazn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109684.1 | NP_001103154.1 | 7 | 1295 | ||
NM_001109685.1 | NP_001103155.1 | 8 | 1313 | |||
NM_144531.3 | NP_653114.3 | 7 | 1295 | |||
GenBank mRNA | AK018497.1 | - | 8 | 1293 | ||
AK032460.1 | - | 5 | 534 | |||
AK052809.1 | - | 8 | 1314 | |||
AK173090.1 | - | 7 | 1295 | |||
BC022941.1 | - | 7 | 1299 | |||
BC100387.1 | - | 5 | 515 |