Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081490.2 | NP_001074959.1 | 8, 9 | 838 | ||
NM_023605.2 | NP_076094.2 | 8, 9 | 1140 | |||
GenBank mRNA | AB041585.1 | - | 8, 9 | 1110 | ||
AK077607.1 | - | 8, 9 | 801 | |||
BC020074.1 | - | 8, 9 | 1111 | 3710611 |