Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027823.1 | NP_082099.1 | 3 | 517 | ||
GenBank mRNA | AK018520.1 | - | 3 | 531 | ||
AK082857.1 | - | 3 | 606 | |||
BC137891.1 | - | 3 | 517 | 8860910 | ||
BC137892.1 | - | 3 | 517 | 8860911 |