Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133733.4 | NP_598494.2 | 6 | 1149 | ||
GenBank mRNA | AB040490.1 | - | 6 | 1066 | ||
AK018527.1 | - | 1 | 140 | |||
AK028983.1 | - | 6 | 1119 | |||
AK033723.1 | - | 6 | 1151 | |||
AK080481.1 | - | 6 | 1105 | |||
AY259213.1 | - | 7 | 778 | |||
AY326421.1 | - | 6 | 784 | |||
BC026447.1 | - | 6 | 975 | 4459717 |