Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027835.3 | NP_082111.2 | 3 | 1047 | ||
GenBank mRNA | AF374384.1 | - | 3 | 956 | ||
AK037057.1 | - | 3 | 945 | |||
AK040519.1 | - | 3 | 1004 | |||
AK149802.1 | - | 3 | 954 | |||
AK153018.1 | - | 3 | 954 | |||
AK156450.1 | - | 3 | 955 | |||
AK168646.1 | - | 3 | 1048 | |||
AY075132.1 | - | 3 | 722 | |||
EF113547.1 | - | 3 | 736 | |||
EF113548.1 | - | 3 | 736 | |||
EU084750.1 | - | 3 | 736 |