Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930506M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114312.1 | NP_001107784.1 | 9 | 1358 | ||
NM_175172.4 | NP_780381.1 | 9 | 1358 | |||
GenBank mRNA | AK015721.1 | - | 2 | 290 | ||
AK039224.1 | - | 5 | 429 | |||
AK082304.1 | - | 9 | 1359 | |||
AK220405.1 | - | 9 | 1388 | |||
BC030338.1 | - | 9 | 930 | 2599101 | ||
EF055486.1 | - | 9 | 801 |