Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002444.2 | NP_002435.1 | 3 | 379 | ||
XM_005262269.2 | XP_005262326.1 | 3 | 400 | |||
XM_011530959.1 | XP_011529261.1 | 3 | 295 | |||
XM_017029545.1 | XP_016885034.1 | 5 | 523 | |||
XM_017029546.1 | XP_016885035.1 | 4 | 419 | |||
GenBank mRNA | AK308486.1 | - | 3 | 342 | ||
BC001112.2 | - | 3 | 351 | |||
BC011827.2 | - | 3 | 351 | |||
BC017293.2 | - | 3 | 332 | 4908580 | ||
CN386296.1 | - | 3 | 379 | |||
EU794636.1 | - | 3 | 332 | |||
M69066.1 | - | 3 | 281 |