Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024243.4 | NP_077205.3 | 3 | 615 | ||
GenBank mRNA | AK002230.1 | - | 3 | 583 | ||
AK053323.1 | - | 3 | 560 | |||
AK089958.1 | - | 3 | 585 | |||
AK150526.1 | - | 3 | 550 | |||
AK150890.1 | - | 3 | 607 | |||
AK151258.1 | - | 3 | 583 | |||
AK151361.1 | - | 3 | 604 | |||
AK151879.1 | - | 3 | 604 | |||
AK151908.1 | - | 3 | 558 | |||
AK152070.1 | - | 3 | 558 | |||
AK165604.1 | - | 3 | 585 | |||
AK170503.1 | - | 3 | 595 | |||
AK201832.1 | - | 2 | 110 | |||
AK204459.1 | - | 2 | 109 | |||
BC003235.1 | - | 3 | 588 | 3497631 |