Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF356878.1 | - | 7 | 850 | ||
AF356879.1 | - | 6 | 709 | |||
AF375479.1 | - | 5 | 661 | |||
AK002247.1 | - | 7 | 849 | |||
AK143782.1 | - | 7 | 852 | |||
AY169234.1 | - | 7 | 849 | |||
BC010795.1 | - | 5 | 1070 | 4221711 |