Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027865.2 | NP_082141.1 | 9 | 1108 | ||
GenBank mRNA | AK002841.1 | - | 9 | 1109 | ||
AK090375.1 | - | 7 | 1382 | |||
AK154206.1 | - | 8 | 1594 | |||
AK165199.1 | - | 9 | 1140 | |||
AK165827.1 | - | 9 | 1093 | |||
BC060057.1 | - | 7 | 1431 | 5696102 | ||
BC060243.1 | - | 7 | 1431 | 5696102 |