Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001609.3 | NP_001600.1 | 8 | 1044 | ||
XM_017016194.1 | XP_016871683.1 | 7 | 885 | |||
GenBank mRNA | AK298638.1 | - | 7 | 873 | ||
AK314241.1 | - | 8 | 940 | |||
AL831821.2 | - | 8 | 913 | |||
BC013756.1 | - | 8 | 939 | 4706281 | ||
KU177879.1 | - | 8 | 927 | |||
KU177880.1 | - | 9 | 1235 | |||
U12778.1 | - | 8 | 942 |