Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023617.2 | NP_076106.2 | 27 | 3226 | ||
GenBank mRNA | AF172276.2 | - | 27 | 3302 | ||
AK004700.1 | - | 16 | 2022 | |||
AK030109.1 | - | 23 | 2766 | |||
AK149434.1 | - | 27 | 3188 | |||
AY347569.1 | - | 27 | 3103 | |||
AY347570.1 | - | 27 | 3103 | |||
BC049853.1 | - | 27 | 3178 | |||
BC138876.1 | - | 27 | 3205 | 8861898 |