Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027890.5 | NP_082166.3 | 3, 4 | 503 | ||
GenBank mRNA | AK004703.2 | - | 3, 4 | 473 | ||
AK028817.1 | - | 3, 4 | 479 | |||
AK037059.1 | - | 3, 4 | 485 | |||
AK037127.1 | - | 3, 4 | 454 | |||
AK038944.1 | - | 3, 4 | 504 | |||
AK042469.1 | - | 3, 4 | 487 | |||
AK128959.1 | - | 3, 4 | 475 | |||
AK167155.1 | - | 3, 4 | 504 | |||
BB632717.1 | - | 3, 4 | 505 | |||
BC026837.1 | - | 3, 4 | 456 | 4222578 | ||
BC066833.1 | - | 1, 2 | 121 |