Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174857.3 | NP_777282.1 | 11 | 1809 | ||
GenBank mRNA | AK004794.1 | - | 10 | 1672 | ||
AK035566.1 | - | 11 | 1810 | |||
AK154219.1 | - | 11 | 1886 | |||
BC042773.1 | - | 11 | 1838 | 4019075 |