Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023626.4 | NP_076115.3 | 7, 8 | 711 | ||
GenBank mRNA | AK030364.1 | - | 6, 7 | 518 | ||
AK080787.1 | - | 7, 8 | 680 | |||
AK131993.1 | - | 7, 8 | 711 | |||
AK146145.1 | - | 6, 7 | 495 | |||
AY007791.1 | - | 7, 8 | 680 |