Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175174.4 | NP_780383.2 | 3 | 767 | ||
GenBank mRNA | AK004982.1 | - | 3 | 503 | ||
AK090246.1 | - | 3 | 327 | |||
AK220195.1 | - | 3 | 513 | |||
BC057606.1 | - | 3 | 767 | 6402367 |