Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175175.4 | NP_780384.1 | 2 | 403 | ||
GenBank mRNA | AK033666.1 | - | 2 | 397 | ||
AK154634.1 | - | 2 | 394 | |||
BC016134.1 | - | 2 | 344 | 4503757 | ||
BC039276.1 | - | 2 | 354 | 4987735 |