Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027934.2 | NP_082210.1 | 6 | 1618 | ||
GenBank mRNA | AK013941.1 | - | 6 | 1484 | ||
AK032259.1 | - | 6 | 1585 | |||
AK044199.1 | - | 3 | 337 | |||
AK136632.1 | - | 6 | 1619 | |||
AK151379.1 | - | 6 | 1593 | |||
AK152404.1 | - | 6 | 1593 | |||
AK157911.1 | - | 1558 | ||||
BC006034.1 | - | 3 | 574 | |||
BC046775.1 | - | 6 | 1639 | 5708557 | ||
BC075700.1 | - | 6 | 1639 | 5708557 |