Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdilt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027943.1 | NP_082219.1 | 6 | 1001 | ||
GenBank mRNA | AK005692.1 | - | 6 | 1001 |