Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027959.3 | NP_082235.1 | 8 | 752 | ||
GenBank mRNA | AK005989.1 | - | 8 | 758 | ||
AK076558.1 | - | 8 | 753 | |||
AK151008.1 | - | 8 | 732 | |||
AK152960.1 | - | 8 | 765 | |||
AK165877.1 | - | 8 | 731 | |||
AK166111.1 | - | 8 | 759 | |||
AK167386.1 | - | 8 | 770 | |||
AK168136.1 | - | 8 | 759 | |||
AK168279.1 | - | 8 | 759 | |||
AK179393.1 | - | 2 | 107 | |||
AK211058.1 | - | 3 | 178 | |||
AK217964.1 | - | 3 | 178 | |||
BC006865.1 | - | 8 | 741 | 2645183 |