Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027973.3 | NP_082249.1 | 9, 10 | 864 | ||
GenBank mRNA | AK006479.1 | - | 9, 10 | 873 | ||
AK050386.1 | - | 9, 10 | 865 | |||
AK081703.1 | - | 9, 10 | 852 | |||
AY366362.1 | - | 5, 6 | 396 | |||
BC094675.1 | - | 10, 11 | 984 | |||
BC117784.2 | - | 9, 10 | 865 | 40092112 | ||
BC117785.2 | - | 9, 10 | 865 | 40092113 |