Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002451.3 | NP_002442.2 | 6 | 743 | ||
GenBank mRNA | AF216650.1 | - | 1 | 87 | ||
AK300592.1 | - | 6 | 906 | |||
AK309365.1 | - | 6 | 730 | |||
AY712791.1 | - | 6 | 751 | |||
BC012316.1 | - | 6 | 717 | 3546198 | ||
BC026106.1 | - | 6 | 725 | 4820938 | ||
CR541670.1 | - | 6 | 630 | |||
CR541710.1 | - | 6 | 630 | |||
FJ224338.1 | - | 6 | 630 | |||
HE654772.1 | - | 6 | 739 | |||
HE654773.1 | - | 6 | 739 | |||
HE654774.1 | - | 6 | 739 | |||
HE654775.1 | - | 6 | 739 | |||
HE654776.1 | - | 6 | 739 | |||
HE654777.1 | - | 6 | 739 | |||
L40432.1 | - | 6 | 740 | |||
U22233.1 | - | 6 | 751 |