Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009587.1 | - | 5 | 687 | ||
AK150216.1 | - | 5 | 680 | |||
AK165627.1 | - | 5 | 491 | |||
BC011282.1 | - | 5 | 617 | 3484522 | ||
BC071264.1 | - | 7 | 617 | 6516319 |