Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam219a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159583.1 | NP_001153055.1 | 2 | 373 | ||
NM_027993.3 | NP_082269.2 | 2 | 340 | |||
GenBank mRNA | AK009522.1 | - | 2 | 324 | ||
AK134737.1 | - | 2 | 374 | |||
BC147808.1 | - | 2 | 302 | 9087525 | ||
BC147810.1 | - | 2 | 300 | 9087527 | ||
BC147834.1 | - | 2 | 302 | 9087551 | ||
BC147836.1 | - | 2 | 302 | 9087553 | ||
BC172167.1 | - | 2 | 269 | 9054837 | ||
BC172173.1 | - | 2 | 302 | 9054843 | ||
BY718591.1 | - | 2 | 374 |