Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285845.1 | NP_001272774.1 | 3 | 679 | ||
NM_001285846.1 | NP_001272775.1 | 4 | 605 | |||
NM_001285847.1 | NP_001272776.1 | 3 | 568 | |||
NM_001285849.1 | NP_001272778.1 | 3 | 452 | |||
NM_027995.3 | NP_082271.1 | 2 | 484 | |||
GenBank mRNA | AA882336.1 | - | 2 | 256 | ||
AF313618.1 | - | 2 | 238 | |||
AK009450.1 | - | 2 | 269 | |||
AK078685.1 | - | 2 | 242 | |||
AK161542.1 | - | 3 | 326 | |||
AK170307.1 | - | 4 | 389 | |||
AY278956.1 | - | 1 | 177 | |||
AY424296.1 | - | 1 | 213 | |||
BC022922.1 | - | 1 | 213 | 4239768 |
Set Membership: |
G-protein coupled receptor |