Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Antxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133738.1 | NP_598499.1 | 5 | 840 | ||
GenBank mRNA | AK155460.1 | - | 5 | 923 | ||
AK166124.1 | - | 612 | ||||
AK170655.1 | - | 5 | 478 | |||
AK171292.1 | - | 5 | 515 | |||
AK190386.1 | - | 5 | 348 | |||
AK193762.1 | - | 2 | 99 | |||
AK207219.1 | - | 5 | 348 | |||
BC076595.1 | - | 5 | 840 | 30649897 |