Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053087.2 | NP_444317.1 | 4 | 316 | ||
GenBank mRNA | AJ291391.1 | - | 4 | 316 | ||
AK010136.1 | - | 3 | 264 | |||
BC119291.1 | - | 4 | 369 | 8734040 | ||
BC119293.1 | - | 4 | 369 | 8734042 | ||
BC145041.1 | - | 4 | 416 |