Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 2, 3 | 287 | ||
GenBank mRNA | AK150361.1 | - | 2, 3 | 235 | ||
AK151044.1 | - | 2, 3 | 255 | |||
AK152180.1 | - | 2, 3 | 235 | |||
AK152423.1 | - | 2, 3 | 200 | |||
AK153122.1 | - | 2, 3 | 200 | |||
AK153539.1 | - | 2, 3 | 235 | |||
AK155767.1 | - | 2, 3 | 255 | |||
AK157001.1 | - | 3, 4 | 212 | |||
AK162610.1 | - | 2, 3 | 283 | |||
AK165671.1 | - | 2, 3 | 254 | |||
AK166444.1 | - | 2, 3 | 200 | |||
AK170823.1 | - | 2, 3 | 246 | |||
AK172352.1 | - | 2, 3 | 216 | |||
AK172553.1 | - | 2, 3 | 216 | |||
AK172701.1 | - | 2, 3 | 216 | |||
AK193384.1 | - | 2, 3 | 77 | |||
AK201658.1 | - | 2, 3 | 78 | |||
BC010292.1 | - | 2, 3 | 236 | 3153174 | ||
BC086319.1 | - | 2, 3 | 244 | 5293409 | ||
BY259389.1 | - | 2, 3 | 288 |