Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 3 | 380 | ||
GenBank mRNA | AK150361.1 | - | 3 | 328 | ||
AK150365.1 | - | 4 | 408 | |||
AK151044.1 | - | 3 | 348 | |||
AK152180.1 | - | 3 | 328 | |||
AK152423.1 | - | 3 | 293 | |||
AK153122.1 | - | 3 | 293 | |||
AK153378.1 | - | 4 | 408 | |||
AK153539.1 | - | 3 | 328 | |||
AK155767.1 | - | 3 | 348 | |||
AK157001.1 | - | 4 | 305 | |||
AK162610.1 | - | 3 | 376 | |||
AK165671.1 | - | 3 | 347 | |||
AK166444.1 | - | 3 | 293 | |||
AK170823.1 | - | 3 | 339 | |||
AK172352.1 | - | 3 | 309 | |||
AK172553.1 | - | 3 | 309 | |||
AK172701.1 | - | 3 | 309 | |||
AK211851.1 | - | 1 | 35 | |||
BC010292.1 | - | 3 | 329 | 3153174 | ||
BC086319.1 | - | 3 | 337 | 5293409 | ||
BY259389.1 | - | 3 | 381 |