Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005956.3 | NP_005947.3 | 27 | 3141 | ||
GenBank mRNA | AK095514.1 | - | 3 | 846 | ||
AK312361.1 | - | 27 | 2846 | |||
BC001014.2 | - | 10 | 1039 | |||
BC009806.2 | - | 27 | 2807 | 3508998 | ||
BC050420.1 | - | 27 | 2832 | 5788176 | ||
J04031.1 | - | 27 | 2807 |