Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028013.3 | NP_082289.2 | 2 | 675 | ||
GenBank mRNA | AK078056.1 | - | 2 | 426 | ||
AK079741.1 | - | 2 | 414 | |||
AK086102.1 | - | 2 | 432 | |||
AK142446.1 | - | 3 | 353 | |||
AK173036.1 | - | 2 | 675 | |||
BC019813.1 | - | 2 | 441 | 4017632 | ||
BC096490.1 | - | 2 | 406 | 6439018 |