Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310067B10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028014.3 | NP_082290.2 | 24 | 3468 | ||
GenBank mRNA | AK047622.1 | - | 10 | 1602 | ||
BC007157.1 | - | 11 | 1714 | |||
BC030865.1 | - | 9 | 1235 | |||
BC053088.1 | - | 24 | 3383 | 5709104 |