Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suds3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122666.2 | NP_001116138.1 | 7, 8 | 804 | ||
NM_178622.5 | NP_848737.3 | 7, 8 | 804 | |||
GenBank mRNA | AF469109.1 | - | 7, 8 | 744 | ||
AK010274.1 | - | 7, 8 | 708 | |||
AK043652.1 | - | 7, 8 | 747 | |||
AK045475.1 | - | 7, 8 | 758 | |||
AK047910.1 | - | 1, 2 | 137 | |||
AK138947.1 | - | 7, 8 | 755 | |||
AK147512.1 | - | 1, 2 | 3812 | |||
AK150633.1 | - | 7, 8 | 742 | |||
BC036562.1 | - | 2, 3 | 177 | |||
BC040088.1 | - | 5, 6 | 417 | |||
BC055764.1 | - | 7, 8 | 757 | 6402735 | ||
BC062176.1 | - | 7, 8 | 671 | 6514171 |