Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005957.4 | NP_005948.3 | 4 | 777 | ||
XM_005263458.3 | XP_005263515.1 | 4 | 1614 | |||
XM_005263460.4 | XP_005263517.1 | 4 | 1232 | |||
XM_005263462.4 | XP_005263519.1 | 4 | 665 | |||
XM_005263463.3 | XP_005263520.1 | 4 | 794 | |||
XM_011541495.2 | XP_011539797.1 | 4 | 1611 | |||
XM_011541496.2 | XP_011539798.1 | 4 | 1614 | |||
XM_017001328.1 | XP_016856817.1 | 4 | 1614 | |||
GenBank mRNA | AB209113.1 | - | 3 | 815 | ||
AJ237672.1 | - | 3 | 677 | |||
AK312907.1 | - | 4 | 638 | |||
BC053509.1 | - | 4 | 599 | 6374885 |