Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 10 | 1590 | ||
NM_001321001.1 | NP_001307930.1 | 9 | 1125 | |||
NM_001321002.1 | NP_001307931.1 | 9 | 1097 | |||
NM_001321003.1 | NP_001307932.1 | 10 | 1307 | |||
NM_001321004.1 | NP_001307933.1 | 8 | 1359 | |||
NM_001321005.1 | NP_001307934.1 | 8 | 1008 | |||
NM_002453.2 | NP_002444.2 | 9 | 1169 | |||
XM_005264335.3 | XP_005264392.1 | 7 | 968 | |||
XM_011532871.2 | XP_011531173.1 | 8 | 1002 | |||
XM_017004162.1 | XP_016859651.1 | 10 | 1298 | |||
XM_017004163.1 | XP_016859652.1 | 9 | 1374 | |||
XM_017004164.1 | XP_016859653.1 | 8 | 986 | |||
XM_017004165.1 | XP_016859654.1 | 9 | 1201 | |||
XM_017004166.1 | XP_016859655.1 | 9 | 1393 | |||
XM_017004167.1 | XP_016859656.1 | 5 | 1470 | |||
GenBank mRNA | AF494407.1 | - | 9 | 1149 | ||
AK225184.1 | - | 9 | 1138 | |||
AK307993.1 | - | 9 | 1149 | |||
BC039848.1 | - | 9 | 1127 | |||
BC064977.1 | - | 9 | 1109 | |||
L34600.1 | - | 9 | 1123 |