Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028037.4 | NP_082313.2 | 7 | 1109 | ||
GenBank mRNA | AK050332.1 | - | 7 | 1313 | ||
AK148195.1 | - | 7 | 1100 | |||
BC027825.1 | - | 7 | 1085 | 5343012 | ||
BC029047.1 | - | 3 | 258 |