Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028042.3 | NP_082318.2 | 9 | 811 | ||
GenBank mRNA | AJ427222.1 | - | 9 | 755 | ||
AK010576.1 | - | 9 | 813 | |||
AK014517.1 | - | 8 | 711 | |||
AK082109.1 | - | 8 | 701 | |||
AK156267.1 | - | 9 | 799 | |||
AK157175.1 | - | 9 | 812 | |||
AK157504.1 | - | 8 | 710 | |||
BC037200.1 | - | 9 | 791 | 3985548 | ||
BC056463.1 | - | 9 | 795 |