Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012335.3 | NP_036467.2 | 4 | 415 | ||
XM_011528024.1 | XP_011526326.1 | 4 | 415 | |||
XM_011528025.1 | XP_011526327.1 | 4 | 415 | |||
XM_011528026.1 | XP_011526328.1 | 4 | 415 | |||
XM_011528027.1 | XP_011526329.1 | 4 | 415 | |||
XM_011528028.2 | XP_011526330.1 | 4 | 415 | |||
XM_017026821.1 | XP_016882310.1 | 4 | 415 | |||
GenBank mRNA | AB210037.1 | - | 4 | 414 | ||
AB290178.1 | - | 4 | 545 | |||
AJ310570.1 | - | 4 | 317 | |||
AK131106.1 | - | 4 | 414 | |||
AK301166.1 | - | 4 | 406 | |||
BC028071.1 | - | 4 | 391 | 5213035 | ||
DA380582.1 | - | 4 | 416 |