Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028049.2 | NP_082325.2 | 3 | 405 | ||
GenBank mRNA | AK002745.1 | - | 3 | 285 | ||
AK005473.1 | - | 2 | 233 | |||
AK181573.1 | - | 2 | 65 | |||
AK181749.1 | - | 2 | 66 | |||
AK184767.1 | - | 2 | 65 | |||
BC018273.1 | - | 3 | 345 | 4458296 |