Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173182.2 | NP_775274.2 | 9, 10 | 1186 | ||
GenBank mRNA | AB098596.1 | - | 9, 10 | 1124 | ||
AK147346.1 | - | 10, 11 | 1197 | |||
AK220297.1 | - | 9, 10 | 1186 | |||
BC067389.1 | - | 9, 10 | 1192 | 30431862 | ||
DQ192037.1 | - | 8, 9 | 1092 |