Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012335.3 | NP_036467.2 | 27 | 3321 | ||
XM_011528024.1 | XP_011526326.1 | 27 | 3384 | |||
XM_011528025.1 | XP_011526327.1 | 27 | 3372 | |||
XM_011528026.1 | XP_011526328.1 | 26 | 3297 | |||
XM_011528027.1 | XP_011526329.1 | 26 | 3249 | |||
XM_017026821.1 | XP_016882310.1 | 27 | 3309 | |||
GenBank mRNA | AB210037.1 | - | 27 | 3320 | ||
AB290178.1 | - | 27 | 3451 | |||
AJ310570.1 | - | 27 | 3223 | |||
AK131106.1 | - | 27 | 3322 | |||
BC028071.1 | - | 27 | 3297 | 5213035 | ||
X98411.1 | - | 18 | 2235 |